Nickel acetate hydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Nickel(II) acetate hydrate
123755-1KG
123755-250G
123755-5KG
BT123755-1KGEA
134
USD
BT123755-1KG
BT123755-250G
BT123755-5KG
Nickel acetate hydrate ≥99%
Nickel acetate hydrate
CAS: 6018-89-9; EC No: 206-761-7; MDL No: MFCD00012451
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: Ni(OCOCH3)2·xH2O; MW: 176.79 (anhydrous)
- Crystalline
Formula:
C₄H₆NiO₄·H₂O MW: 194.79 g/mol |
MDL Number:
MFCD00012451 CAS Number: 6018-89-9 EINECS: 206-761-7 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...