Strontium ≥99% (trace metals basis), granules ≤20 mm

Supplier: BeanTown Chemical

123665-100G 123665-25G 123665-5G
BT123665-100GEA 519 USD
BT123665-100G BT123665-25G BT123665-5G
Strontium ≥99% (trace metals basis), granules ≤20 mm
Strontium

CAS: 7440-24-6; EC No: 231-133-4; MDL No: MFCD00134060; RTECS: WK7700000 UN No: UN3208; Haz Class: 4.3; Packing Group: II Solid, packaged under argon with thin oil layer; Molecular Formula: Sr; MW: 87.62 Melting Point: 757°; Boiling Point: 1384° Density (g/mL): 2.6

Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1384 °C
Melting Pt: 757 °C
Density: 2.6 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR