Niobium isopropoxide ≥99%

Supplier: BeanTown Chemical

123660-100ML 123660-25ML
BT123660-100MLEA 420 USD
BT123660-100ML BT123660-25ML
Niobium isopropoxide ≥99%
Niobium isopropoxide

CAS: 18368-80-4; MDL No: MFCD00144915 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: Nb[OCH(CH3)2]5; MW: 388.35 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₁₅H₃₅NbO₅
MW: 388.346 g/mol
MDL Number: MFCD00144915
CAS Number: 18368-80-4
ADR: 3 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR