1-Boc-piperazine ≥99%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

123590-1G 123590-25G 123590-5G
BT123590-1GEA 31.21 USD
BT123590-1G BT123590-25G BT123590-5G
1-Boc-piperazine ≥99%
1-Boc-piperazine

CAS: 57260-71-6; MDL No: MFCD00075265 UN No: UN3259; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C9H18N2O2; MW: 186.25 Melting Point: 43-47° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₉H₁₈N₂O₂
Melting Pt: 43…47 °C
MDL Number: MFCD00075265
CAS Number: 57260-71-6
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR