(3-Aminopropyl)triethoxysilane ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane

123580-100G 123580-2.5KG 123580-500G
BT123580-100GEA 45.07 USD
BT123580-100G BT123580-2.5KG BT123580-500G
(3-Aminopropyl)triethoxysilane ≥98%
(3-Aminopropyl)triethoxysilane

CAS: 919-30-2; EC No: 213-048-4; MDL No: MFCD00008207; RTECS: TX2100000 UN No: UN2735; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C9H23NO3Si; MW: 221.37 Boiling Point: 217°; Flash point: 90°C (194°F) Density (g/mL): 0.946; Refractive Index: 1.421 Air Sensitive, Moisture Sensitive


  • RTECS: TX2100000
  • Refractive index: 1.421
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217 °C
Density: 0.946 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR