Pivalaldehyde (trimethylacetaldehyde) ≥96%

Supplier: BeanTown Chemical

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

123560-100G 123560-25G 123560-5G
BT123560-100GEA 644 USD
BT123560-100G BT123560-25G BT123560-5G
Pivalaldehyde (trimethylacetaldehyde) ≥96%
Pivalaldehyde (trimethylacetaldehyde)

CAS: 630-19-3; EC No: 211-134-6; MDL No: MFCD00006962 UN No: UN1989; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CCHO; Molecular Formula: C5H10O; MW: 86.13 Melting Point: 6°; Boiling Point: 74°; Flash point: -16°C (3°F) Density (g/mL): 0.793; Refractive Index: 1.378 Air Sensitive


  • Refractive index: 1.378
  • Air Sensitive
  • Liquid

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 74 °C
Melting Pt: 6 °C
Density: 0.793 g/mL
Flash Pt: –16 °C (3 °F)
MDL Number: MFCD00006962
CAS Number: 630-19-3
EINECS: 211-134-6
ADR: 3,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR