Arsenic(III) oxide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
di-Arsenic trioxide
123535-25G
123535-5G
BT123535-25GEA
134.63
USD
BT123535-25G
BT123535-5G
Arsenic(III) oxide ≥99.995%
Arsenic(III) oxide
CAS: 1327-53-3; EC No: 215-481-4; MDL No: MFCD00003433; RTECS: CG3325000
UN No: UN1561; Haz Class: 6.1; Packing Group: II
Powder; Molecular Formula: As2O3; MW: 197.84
Boiling Point: 465° (sublimes)
- RTECS: CG3325000
- Powder
Formula:
As₂O₃ MW: 197.84 g/mol Boiling Pt: 465 (subl.) Melting Pt: 312 |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1 II Merck Index: 12,00844 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...