Potassium bis(trimethylsilyl)amide 20% (w/w) 0.91 M in THF

Supplier: BeanTown Chemical

123475-100ML 123475-25ML
BT123475-100MLEA 263.97 USD
BT123475-100ML BT123475-25ML
Potassium bis(trimethylsilyl)amide 20% (w/w) 0.91 M in THF
Potassium bis(trimethylsilyl)amide

CAS: 40949-94-8; MDL No: MFCD00010330 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: [(CH3)3Si]2NK; Molecular Formula: C6H18KNSi2; MW: 199.49 Boiling Point: 66°; Flash point: -18°C (-2°F) Density (g/mL): 0.915 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₆H₁₈KNSi₂
Boiling Pt: 66 °C
Density: 0.915 g/mL
Flash Pt: –18 °C (–2 °F)
MDL Number: MFCD00010330
CAS Number: 40949-94-8
ADR: 3(8),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR