Silver(I) acetate, anhydrous ≥99%

Supplier: BeanTown Chemical

123430-100G 123430-25G
BT123430-100GEA 380.6 USD
BT123430-100G BT123430-25G
Silver(I) acetate, anhydrous ≥99%
Silver(I) acetate

CAS: 563-63-3; EC No: 209-254-9; MDL No: MFCD00012446; RTECS: AJ4100000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Powder; Linear Formula: AgOOCCH3; MW: 166.92 Density (g/mL): 3.259 Light Sensitive


  • RTECS: AJ4100000
  • Light Sensitive
  • Crystalline/Powder

Formula: C₂H₃AgO₂
Density: 3.259 g/mL
MDL Number: MFCD00012446
CAS Number: 563-63-3
EINECS: 209-254-9
ADR: 9,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR