Primaquine diphosphate ≥98%
Supplier: BeanTown Chemical
Synonyms:
N4-(6-Methoxyquinolin-8-yl)pentane-1,4-diamine bis(phosphate), NSC 149765, Primaquine bisphosphate
123415-10G
123415-1G
BT123415-10GEA
189
USD
BT123415-10G
BT123415-1G
Primaquine diphosphate ≥98%
Primaquine diphosphate
CAS: 63-45-6; EC No: 200-560-8; MDL No: MFCD00013166; RTECS: VA9660000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: C15H21N3O·2H3PO4; Molecular Formula: C15H21N3O·2H3O4P; MW: 455.34
Melting Point: 205-206° (decomposes)
- RTECS: VA9660000
- Powder
Formula:
C₁₅H₂₇N₃O₉P₂ MW: 455.33 g/mol Melting Pt: 205…206 °C (dec.) |
MDL Number:
MFCD00013166 CAS Number: 63-45-6 EINECS: 200-560-8 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...