Ruthenium(III) bromide hydrate

Supplier: BeanTown Chemical

123375-1G 123375-5G
BT123375-1GEA 67 USD
BT123375-1G BT123375-5G
Ruthenium(III) bromide hydrate
Ruthenium(III) bromide hydrate

CAS: 14014-88-1; EC No: 237-829-4; MDL No: MFCD00016314 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: RuBr3·xH2O; MW: 340.78 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: RuBr₃·xH₂O
MW: 340.80 g/mol (anhy)
MDL Number: MFCD00016314
CAS Number: 314075-07-5
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR