n-Octane-[D₁₈] ≥99% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: n-Octane-[²H₁₈], Octane-D18

123335-1G 123335-5G
BT123335-1GEA 213 USD
BT123335-1G BT123335-5G
n-Octane-[D₁₈] ≥99% (Isotopic)
n-Octane-[D₁₈]

CAS: 17252-77-6; EC No: 241-285-3; MDL No: MFCD00037626 UN No: UN1262; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CD3(CD2)6CD3 ; Molecular Formula: C8D18; MW: 132.34 Melting Point: -57°; Boiling Point: 125-127°; Flash point: 16°C (61°F) Density (g/mL): 0.815; Refractive Index: 1.394 Hygroscopic


  • Refractive index: 1.394
  • Hygroscopic
  • Liquid

Formula: D₃C(CD₂)₆CD₃
MW: 132.09 g/mol
Boiling Pt: 125…127 °C
Melting Pt: –57 °C
Density: 0.815 g/mL
Flash Pt: 16 °C (61 °F)
MDL Number: MFCD00037626
CAS Number: 17252-77-6
EINECS: 241-285-3
UN: 1262
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR