Cyclohexene ≥99%
Supplier: BeanTown Chemical
123275-100ML
123275-2.5L
BT123275-100MLEA
29.57
USD
BT123275-100ML
BT123275-2.5L
Cyclohexene ≥99%
Cyclohexene
CAS: 110-83-8; EC No: 203-807-8; MDL No: MFCD00001539; RTECS: GW2500000
UN No: UN2256; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H10; MW: 82.15
Melting Point: -104°; Boiling Point: 83°; Flash point: -12°C (10°F)
Density (g/mL): 0.811; Refractive Index: 1.446
- RTECS: GW2500000
- Refractive index: 1.446
- Liquid
Formula:
C₆H₁₀ MW: 82.15 g/mol Boiling Pt: 83 °C Melting Pt: –104 °C Density: 0.811 g/mL Flash Pt: –12 °C (10 °F) |
MDL Number:
MFCD00001539 CAS Number: 110-83-8 EINECS: 203-807-8 UN: 2256 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...