2-Butynoic acid ≥98%

Supplier: BeanTown Chemical

123250-1G 123250-25G 123250-5G
BT123250-1GEA 50 USD
BT123250-1G BT123250-25G BT123250-5G
2-Butynoic acid ≥98%
2-Butynoic acid

CAS: 590-93-2; EC No: 209-695-7; MDL No: MFCD00004363 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Molecular Formula: C4H4O2; MW: 84.07 Melting Point: 78-80°


  • Powder

Formula: C₄H₄O₂
MW: 84.07 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR