Aniline ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Aminobenzene, Phenylamine
123230-100ML
123230-1L
123230-500ML
BT123230-100MLEA
31.6
USD
BT123230-100ML
BT123230-1L
BT123230-500ML
Aniline ≥99.0% ACS
Aniline
CAS: 62-53-3; EC No: 200-539-3; MDL No: MFCD00007629; RTECS: BW6650000
UN No: UN1547; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C6H7N; MW: 93.13
Melting Point: -6°; Boiling Point: 184°; Flash point: 70°C (158°F)
Density (g/mL): 1.022; Refractive Index: 1.586
- RTECS: BW6650000
- Refractive index: 1.586
- Liquid
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 184 °C Melting Pt: –6 °C Density: 1.022 g/mL Flash Pt: 70 °C (158 °F) |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1,II |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...