Zinc sulphate 100 mM in water

Supplier: BeanTown Chemical

123220-100ML 123220-50ML
BT123220-100MLEA 54.83 USD
BT123220-100ML BT123220-50ML
Zinc sulphate 100 mM in water
Zinc sulphate

CAS: 7733-02-0; EC No: 231-793-3; MDL No: MFCD00011302; RTECS: ZH5260000 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Linear Formula: ZnSO4; MW: 161.45 Density (g/mL): 1.31


  • RTECS: ZH5260000
  • Liquid

Formula: O₄SZn
MW: 161.45
Melting Pt: 100
Density: 1.31 g/mL
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9 III
Merck Index: 12,10293

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR