Calcium sulfide ≥99% (trace metals basis excluding Mg)

Supplier: BeanTown Chemical

123185-100G 123185-25G
BT123185-100GEA 628.62 USD
BT123185-100G BT123185-25G
Calcium sulfide ≥99% (trace metals basis excluding Mg)
Calcium sulphide

CAS: 20548-54-3; EC No: 243-873-5; MDL No: MFCD00015990 UN No: UN3134; Haz Class: 4.3 (6.1); Packing Group: III -325 Mesh Powder; Molecular Formula: CaS; MW: 72.14 Moisture Sensitive


  • Moisture Sensitive
  • -325 Mesh Powder

Formula: CaS
MW: 72.14 g/mol
MDL Number: MFCD00015990
CAS Number: 20548-54-3
EINECS: 243-873-5
UN: 3134
ADR: 4.3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR