4-(Dimethylamino)antipyrine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,3-Dimethyl-4-(dimethylamino)-1-phenyl-3-pyrazolin-5-one, 4-(Dimethylamino)phenazone

123175-1KG 123175-250G 123175-50G
BT123175-1KGEA 586 USD
BT123175-1KG BT123175-250G BT123175-50G
4-(Dimethylamino)antipyrine ≥98%
4-(Dimethylamino)antipyrine

CAS: 58-15-1; MDL No: MFCD00003142; RTECS: CD2625000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C13H17N3O; MW: 231.29 Melting Point: 107-109° Air Sensitive, Light Sensitive


  • RTECS: CD2625000
  • Air Sensitive, Light Sensitive
  • Crystalline/Powder

Formula: C₁₃H₁₇N₃O
MW: 231.3 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD00003142
CAS Number: 58-15-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR