3-Amino-2-methylbenzotrifluoride ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Methyl-3-(trifluoromethyl)aniline

123155-1G 123155-5G
BT123155-1GEA 81 USD
BT123155-1G BT123155-5G
3-Amino-2-methylbenzotrifluoride ≥99%
3-Amino-2-methylbenzotrifluoride

CAS: 54396-44-0; EC No: 259-145-5; MDL No: MFCD00153216 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C8H8F3N ; MW: 175.15 Melting Point: 38-42°; Flash point: 96°C (205°F)


  • Solid

Formula: C₈H₈F₃N
MW: 175.16 g/mol
Melting Pt: 38…42 °C
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00153216
CAS Number: 54396-44-0
EINECS: 259-145-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR