Hydrazinium monobromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Hydrazine monohydrobromide
123115-250G
123115-50G
BT123115-250GEA
124
USD
BT123115-250G
BT123115-50G
Hydrazinium monobromide ≥98%
Hydrazinium monobromide
CAS: 13775-80-9; EC No: 237-412-7; MDL No: MFCD00065394
UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II
Solid; Linear Formula: H2NNH2·HBr ; MW: 112.96
Melting Point: 87-92°
Hygroscopic
- Hygroscopic
- Solid
Formula:
H₅BrN₂ Melting Pt: 87…92 °C |
MDL Number:
MFCD00065394 CAS Number: 13775-80-9 EINECS: 237-412-7 UN: 2928 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...