4-Chlorobenzylamine ≥97%

Supplier: BeanTown Chemical

Synonyms: (4-Chlorophenyl)methanamine

123060-100G 123060-25G 123060-5G
BT123060-100GEA 225 USD
BT123060-100G BT123060-25G BT123060-5G
4-Chlorobenzylamine ≥97%
4-Chlorobenzylamine

CAS: 104-86-9; EC No: 203-245-3; MDL No: MFCD00008121 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H8ClN; MW: 141.60 Boiling Point: 215°; Flash point: 90°C (194°F) Density (g/mL): 1.164; Refractive Index: 1.558 Air Sensitive


  • Refractive index: 1.558
  • Air Sensitive
  • Liquid

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215 °C
Density: 1.164 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00008121
CAS Number: 104-86-9
EINECS: 203-245-3
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR