Tetramethylammonium perchlorate
Supplier: BeanTown Chemical
123050-100G
123050-25G
BT123050-100GEA
305
USD
BT123050-100G
BT123050-25G
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate
CAS: 2537-36-2; EC No: 219-805-5; MDL No: MFCD00043207
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: (CH3)4N(ClO4); Molecular Formula: C4H12ClNO4; MW: 173.60
Melting Point:
- Crystalline
Formula:
C₄H₁₂ClNO₄ Melting Pt: >300 °C |
MDL Number:
MFCD00043207 CAS Number: 2537-36-2 EINECS: 219-805-5 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...