Bis(benzonitrile)palladium(II) dichloride

Supplier: BeanTown Chemical

Synonyms: Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride

122975-1G 122975-500MG
BT122975-1GEA 114 USD
BT122975-1G BT122975-500MG
Bis(benzonitrile)palladium(II) dichloride
Bis(benzonitrile)dichloropalladium(II)

CAS: 14220-64-5; EC No: 238-085-3; MDL No: MFCD00013123 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C14H10Cl2N2Pd; MW: 383.57 Melting Point: 131°


  • Powder

Formula: C₁₄H₁₀Cl₂N₂Pd
MW: 383.57 g/mol
Melting Pt: 131 °C
MDL Number: MFCD00013123
CAS Number: 14220-64-5
EINECS: 238-085-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR