Borane dimethyl sulfide complex ≥94%

Supplier: BeanTown Chemical

Synonyms: Borane-methyl sulphide complex

122925-100ML 122925-25ML
BT122925-100MLEA 268.21 USD
BT122925-100ML BT122925-25ML
Borane dimethyl sulfide complex ≥94%
Borane dimethyl sulphide complex

CAS: 13292-87-0; EC No: 236-313-6; MDL No: MFCD00013189; RTECS: PV5080000 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: (CH3)2S·BH3; Molecular Formula: C2H9BS; MW: 75.97 Flash point: 18°C (64°F) Density (g/mL): 0.801 Air Sensitive, Moisture Sensitive


  • RTECS: PV5080000
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₂H₉BS
MW: 75.97 g/mol
Density: 0.801 g/mL
Flash Pt: 18 °C (64 °F)
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR