Copper(II) chloride, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Copper dichloride, Cupric chloride

122905-1KG 122905-250G 122905-50G
BT122905-1KGEA 115 USD
BT122905-1KG BT122905-250G BT122905-50G
Copper(II) chloride, anhydrous ≥98%
Copper(II) chloride

CAS: 7447-39-4; EC No: 231-210-2; MDL No: MFCD00010972; RTECS: GL7000000 UN No: UN2802; Haz Class: 8; Packing Group: III Powder; Linear Formula: CuCl2; MW: 134.45 Melting Point: 620° Density (g/mL): 3.386 Air Sensitive , Hygroscopic


  • RTECS: GL7000000
  • Air Sensitive, Hygroscopic
  • Powder

Formula: Cl₂Cu
MW: 134.45 g/mol
Melting Pt: 620 °C
Density: 3.386 g/mL
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR