Acetic anhydride ≥97% ACS

Supplier: BeanTown Chemical

122890-100ML 122890-2L 122890-500ML
BT122890-100MLEA 22.93 USD
BT122890-100ML BT122890-2L BT122890-500ML
Acetic anhydride ≥97% ACS
Acetic anhydride

CAS: 108-24-7; EC No: 203-564-8; MDL No: MFCD00008705; RTECS: AK1925000 UN No: UN1715; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: (CH3CO)2O; MW: 102.09 Melting Point: -73°; Boiling Point: 138-140°; Flash point: 49°C (120°F) Density (g/mL): 1.08; Refractive Index: 1.390 Moisture Sensitive


  • RTECS: AK1925000
  • Refractive index: 1.390
  • Moisture Sensitive
  • Liquid

Formula: (CH₃CO)₂O
Boiling Pt: 138…140 °C
Melting Pt: –73 °C
Density: 1.08 g/mL
Flash Pt: 49 °C (120 °F)
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97% min.
Color (APHA) ≤10
Residue after evaporation ≤0.003%
Chloride (Cl) ≤5ppm
Heavy metals (as Pb) ≤2ppm
Iron (Fe) ≤5ppm
Phosphate (PO4) ≤0.001%
Sulfate (SO4) ≤5ppm
Substances reducing KMnO4 Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR