Lithium tetrafluoroborate ≥98%

Supplier: BeanTown Chemical

122880-10G 122880-50G
BT122880-10GEA 53.5 USD
BT122880-10G BT122880-50G
Lithium tetrafluoroborate ≥98%
Lithium tetrafluoroborate

CAS: 14283-07-9; EC No: 238-178-9; MDL No: MFCD00011087 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: LiBF4; MW: 93.75 Melting Point: 293-300° (decomposes) Hygroscopic


  • Hygroscopic
  • Powder

Formula: BF₄Li
MW: 93.74 g/mol
Melting Pt: 293…300 °C (dec.)
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR