Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate

122840-100G 122840-25G 122840-500G
BT122840-100GEA 102.62 USD
BT122840-100G BT122840-25G BT122840-500G
Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS
Cobalt(II) acetate tetrahydrate

CAS: 6147-53-1; EC No: 200-755-8; MDL No: MFCD00149569; RTECS: AG3325000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Linear Formula: (CH3CO2)2Co·4H2O; Molecular Formula: C4H6CoO4·4H2O; MW: 249.08 Hygroscopic


  • RTECS: AG3325000
  • Hygroscopic
  • Crystalline

Formula: (H₃CCOO)₂Co·4H₂O
MW: 249.08 g/mol
MDL Number: MFCD00149569
CAS Number: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98.0-102.0%
Insoluble matter ≤0.01%
Chloride (Cl) ≤0.002%
Nitrate (NO3) ≤0.01%
Sulfate (SO4) ≤0.005%
Calcium (Ca) ≤0.005%
Copper (Cu) ≤0.002%
Iron (Fe) ≤0.001%
Lead (Pb) ≤0.001%
Magnesium (Mg) ≤0.005%
Nickel (Ni) ≤0.1%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.05%
Zinc (Zn) ≤0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR