2-Cyano-6-methylpyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Methylpicolinonitrile, 6-Methyl-2-pyridinecarbonitrile, 6-Cyano-2-picoline, 6-Methylpyridine-2-carbonitrile

122830-25G 122830-5G
BT122830-25GEA 165 USD
BT122830-25G BT122830-5G
2-Cyano-6-methylpyridine ≥98%
2-Cyano-6-methylpyridine

CAS: 1620-75-3; EC No: 216-588-9; MDL No: MFCD00190586 UN No: UN3439; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: NCC6H4CH3; Molecular Formula: C7H6N2; MW: 118.14 Melting Point: 70-74°


  • Solid

Formula: C₇H₆N₂
MW: 118.14 g/mol
Melting Pt: 70…74 °C
MDL Number: MFCD00190586
CAS Number: 1620-75-3
EINECS: 216-588-9
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR