6-Chloronicotinoyl chloride ≥98%

Supplier: BeanTown Chemical

122770-100G 122770-25G 122770-5G
BT122770-100GEA 1071.68 USD
BT122770-100G BT122770-25G BT122770-5G
6-Chloronicotinoyl chloride ≥98%
6-Chloronicotinoyl chloride

CAS: 58757-38-3; MDL No: MFCD00051775 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline/Powder; Molecular Formula: C6H3Cl2NO; MW: 176.00 Melting Point: 48-53° Moisture Sensitive


  • Moisture Sensitive
  • Crystalline/Powder

Formula: C₆H₃Cl₂NO
Melting Pt: 48…53 °C
MDL Number: MFCD00051775
CAS Number: 58757-38-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR