Tetraethylammonium perchlorate ≥98% (dry weight)
Supplier: BeanTown Chemical
122750-10G
122750-50G
BT122750-10GEA
57
USD
BT122750-10G
BT122750-50G
Tetraethylammonium perchlorate ≥98% (dry weight)
Tetraethylammonium perchlorate
CAS: 2567-83-1; EC No: 219-904-3; MDL No: MFCD00041977
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Wet Crystalline; Linear Formula: (CH3CH2)4N(ClO4); Molecular Formula: C8H20ClNO4; MW: 229.71
Melting Point:
- Wet Crystalline
Formula:
C₈H₂₀ClNO₄ MW: 229.7 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00041977 CAS Number: 2567-83-1 EINECS: 219-904-3 UN: 1479 ADR: 5.1 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...