2-Butoxyethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

122745-10L 122745-2.5L 122745-500ML
BT122745-10LEA 193 USD
BT122745-10L BT122745-2.5L BT122745-500ML
2-Butoxyethanol ≥99%
2-Butoxyethanol

CAS: 111-76-2; EC No: 203-905-0; MDL No: MFCD00002884; RTECS: KJ8575000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H14O2; MW: 118.17 Melting Point: -75°; Boiling Point: 169-173°; Flash point: 67°C (153°F) Density (g/mL): 0.902; Refractive Index: 1.419


  • RTECS: KJ8575000
  • Refractive index: 1.419
  • Liquid

Formula: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Boiling Pt: 169…173 °C
Melting Pt: –75 °C
Density: 0.902 g/mL
Flash Pt: 67 °C (153 °F)
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR