Lithium perchlorate, anhydrous ≥95.0% ACS
Supplier: BeanTown Chemical
122680-100G
122680-500G
BT122680-100GEA
71.51
USD
BT122680-100G
BT122680-500G
Lithium perchlorate, anhydrous ≥95.0% ACS
Lithium perchlorate
CAS: 7791-03-9; EC No: 232-237-2; MDL No: MFCD00011079
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: LiClO4; MW: 106.40
Melting Point: 236°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
ClLiO₄ MW: 106.39 g/mol Melting Pt: 236 °C |
MDL Number:
MFCD00011079 CAS Number: 7791-03-9 EINECS: 232-237-2 UN: 1481 ADR: 5.1,II |
Specification Test Results
Assay | 95.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...