1-Chloro-2-methylpropane ≥98%

Supplier: BeanTown Chemical

122670-250G 122670-50G
BT122670-250GEA 150.4 USD
BT122670-250G BT122670-50G
1-Chloro-2-methylpropane ≥98%
1-Chloro-2-methylpropane

CAS: 513-36-0; EC No: 208-157-9; MDL No: MFCD00000954 UN No: UN1127; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCH2Cl; Molecular Formula: C4H9Cl; MW: 92.57 Melting Point: -131°; Boiling Point: 68-69°; Flash point: -19°C (-2°F) Density (g/mL): 0.883; Refractive Index: 1.398


  • Refractive index: 1.398
  • Liquid

Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68…69 °C
Melting Pt: –131 °C
Density: 0.883 g/mL
Flash Pt: –19 °C (–2 °F)
MDL Number: MFCD00000954
CAS Number: 513-36-0
EINECS: 208-157-9
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR