4-Fluoro-2-(trifluoromethyl)benzoyl chloride ≥97%
Supplier: BeanTown Chemical
122615-1G
122615-5G
BT122615-1GEA
44
USD
BT122615-1G
BT122615-5G
4-Fluoro-2-(trifluoromethyl)benzoyl chloride ≥97%
4-Fluoro-2-(trifluoromethyl)benzoyl chloride
CAS: 189807-21-4; MDL No: MFCD00061158
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C8H3ClF4O; MW: 226.56
Boiling Point: 157°; Flash point: 85°C (185°F)
Density (g/mL): 1.495; Refractive Index: 1.468
Moisture Sensitive
- Refractive index: 1.468
- Moisture Sensitive
- Liquid
Formula:
C₈H₃ClF₄O Boiling Pt: 157 °C Density: 1.495 g/mL Flash Pt: 85 °C (185 °F) |
MDL Number:
MFCD00061158 CAS Number: 189807-21-4 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...