Zinc sulfate heptahydrate 99.0-103.0% ACS

Supplier: BeanTown Chemical

122545-100G 122545-2.5KG 122545-500G
BT122545-100GEA 36.73 USD
BT122545-100G BT122545-2.5KG BT122545-500G
Zinc sulfate heptahydrate 99.0-103.0% ACS
Zinc sulphate heptahydrate

CAS: 7446-20-0; EC No: 231-793-3; MDL No: MFCD00149894; RTECS: ZH5300000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Lump; Linear Formula: ZnSO4·7H2O ; MW: 287.56 Density (g/mL): 1.957


  • RTECS: ZH5300000
  • Crystalline/Lump

Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Density: 1.957 g/mL
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.0-103.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR