1-Bromo-3-methyl-2-butene ≥90% stabilized, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene

122480-10G 122480-250G 122480-50G
BT122480-10GEA 37.55 USD
BT122480-10G BT122480-250G BT122480-50G
1-Bromo-3-methyl-2-butene ≥90% stabilized, Technical Grade
1-Bromo-3-methyl-2-butene

CAS: 870-63-3; EC No: 212-799-5; MDL No: MFCD00000242 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: (CH3)2C=CHCH2Br; Molecular Formula: C5H9Br; MW: 149.03 Boiling Point: 82-83°/150 mmHg; Flash point: 40°C (104°F) Density (g/mL): 1.29; Refractive Index: 1.489 Light Sensitive


  • Refractive index: 1.489
  • Light Sensitive
  • Liquid

Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 82…83 °C/150 mmHg
Density: 1.29 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00000242
CAS Number: 870-63-3
EINECS: 212-799-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR