Cyclobutanol ≥96%

Supplier: BeanTown Chemical

122455-1G 122455-5G
BT122455-1GEA 114 USD
BT122455-1G BT122455-5G
Cyclobutanol ≥96%
Cyclobutanol

CAS: 2919-23-5; EC No: 220-858-1; MDL No: MFCD00001318 UN No: UN1120; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8O; MW: 72.11 Boiling Point: 123°/733 mmHg; Flash point: 31°C (70°F) Density (g/mL): 0.921; Refractive Index: 1.435


  • Refractive index: 1.435
  • Liquid

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 123 °C/733 mmHg
Density: 0.921 g/mL
Flash Pt: 31 °C (70 °F)
MDL Number: MFCD00001318
CAS Number: 2919-23-5
EINECS: 220-858-1
UN: 1120
ADR: 3,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR