Pentafluorophenyl isothiocyanate ≥96%

Supplier: BeanTown Chemical

122440-1G 122440-5G
BT122440-1GEA 84 USD
BT122440-1G BT122440-5G
Pentafluorophenyl isothiocyanate ≥96%
Pentafluorophenyl isothiocyanate

CAS: 35923-79-6; EC No: 252-791-9; MDL No: MFCD00041042 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III Liquid; Molecular Formula: C7F5NS; MW: 225.14 Boiling Point: 109-110°/35 mmHg Density (g/mL): 1.594; Refractive Index: 1.540 Moisture Sensitive


  • Refractive index: 1.540
  • Moisture Sensitive
  • Liquid

Formula: C₇F₅NS
Boiling Pt: 109…110 °C/35 mmHg
Density: 1.594 g/mL
MDL Number: MFCD00041042
CAS Number: 35923-79-6
EINECS: 252-791-9
ADR: 8 (6.1),III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR