1,2,4,5-Tetramethylbenzene ≥97%
Supplier: BeanTown Chemical
122430-100G
122430-500G
BT122430-100GEA
24
USD
BT122430-100G
BT122430-500G
1,2,4,5-Tetramethylbenzene ≥97%
1,2,4,5-Tetramethylbenzene
CAS: 95-93-2; EC No: 202-465-7; MDL No: MFCD00008528; RTECS: DC0500000
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Crystalline; Linear Formula: C6H2(CH3)4; Molecular Formula: C10H14; MW: 134.22
Melting Point: 76-90°; Boiling Point: 196-197°; Flash point: 74°C (165°F)
Density (g/mL): 0.838
- RTECS: DC0500000
- Crystalline
Formula:
C₁₀H₁₄ MW: 134.22 g/mol Boiling Pt: 196…197 °C Melting Pt: 76…90 °C Density: 0.838 g/mL Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00008528 CAS Number: 95-93-2 EINECS: 202-465-7 UN: 1325 ADR: 4.1,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...