Copper(I) chloride ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cuprous chloride, Copper chloride
122405-100G
122405-25G
122405-5G
BT122405-100GEA
491
USD
BT122405-100G
BT122405-25G
BT122405-5G
Copper(I) chloride ≥99.999% (trace metals basis)
Copper(I) chloride
CAS: 7758-89-6; EC No: 231-842-9; MDL No: MFCD00010971; RTECS: GL6990000
UN No: UN2802; Haz Class: 8; Packing Group: III
Powder; Linear Formula: CuCl; MW: 98.99
Melting Point: 430°; Boiling Point: 1490°
Density (g/mL): 4.14
Air Sensitive, Moisture Sensitive
- RTECS: GL6990000
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
ClCu MW: 99 g/mol Boiling Pt: 1490 °C Melting Pt: 430 °C Density: 4.14 g/mL |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8,III |
Specification Test Results
Assay | 99.999% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...