4-Amino-2-chloropyrimidine-5-carbonitrile ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Amino-2-chloro-5-pyrimidinecarbonitrile
122400-10G
122400-2G
122400-500MG
BT122400-10GEA
510
USD
BT122400-10G
BT122400-2G
BT122400-500MG
4-Amino-2-chloropyrimidine-5-carbonitrile ≥97%
4-Amino-2-chloropyrimidine-5-carbonitrile
CAS: 94741-69-2; MDL No: MFCD00052343
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Powder/Flakes; Molecular Formula: C5H3ClN4; MW: 154.56
Melting Point: 200° (decomposes)
- Powder/Flakes
Formula:
C₅H₃ClN₄ MW: 154.56 g/mol Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00052343 CAS Number: 94741-69-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...