Gallium tribromide, anhydrous

Supplier: BeanTown Chemical

Synonyms: Gallium (III) bromide

122375-1G 122375-5G
BT122375-1GEA 68.79 USD
BT122375-1G BT122375-5G
Gallium tribromide, anhydrous
Gallium tribromide

CAS: 13450-88-9; EC No: 236-609-5; MDL No: MFCD00016100 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: GaBr3; MW: 309.45 Melting Point: 122°; Boiling Point: 279° Density (g/mL): 3.69 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: Br₃Ga
MW: 309.44 g/mol
Boiling Pt: 279 °C
Melting Pt: 122 °C
Density: 3.69 g/mL
MDL Number: MFCD00016100
CAS Number: 13450-88-9
EINECS: 236-609-5
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR