Indium(III) isopropoxide 5% (w/v) in 2-propanol , 99% (trace metals basis)
Supplier: BeanTown Chemical
122315-100ML
122315-25ML
BT122315-100MLEA
310
USD
BT122315-100ML
BT122315-25ML
Indium(III) isopropoxide 5% (w/v) in 2-propanol , 99% (trace metals basis)
Indium (III) isopropoxide
CAS: 38218-24-5; MDL No: MFCD00210318
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: In(OCH(CH3)2)3; MW: 292.08
Boiling Point: 83-87°; Flash point: 12°C (54°F)
Density (g/mL): 0.808
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₉H₂₁InO₃ MW: 292.08 g/mol Boiling Pt: 83…87 °C Density: 0.808 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00210318 CAS Number: 38218-24-5 UN: 1219 ADR: 4.1 II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...