α-Bromo-4-(difluoromethoxy)toluene ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-(Bromomethyl)-4-(difluoromethoxy)benzene, p-(Difluoromethoxy)benzyl bromide, 4-(Difluoromethoxy)benzyl bromide

122270-1G 122270-5G
BT122270-1GEA 112 USD
BT122270-1G BT122270-5G
α-Bromo-4-(difluoromethoxy)toluene ≥97%
α-Bromo-4-(difluoromethoxy)toluene

CAS: 3447-53-8; MDL No: MFCD00085007 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C8H7BrF2O ; MW: 237.04 Boiling Point: 64-66°/0.3 mmHg


  • Liquid

Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Boiling Pt: 64…66 °C/0.3 mmHg
MDL Number: MFCD00085007
CAS Number: 3447-53-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR