Methyl ethyl ketone ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

122260-1L 122260-4L 122260-500ML
BT122260-1LEA 70.44 USD
BT122260-1L BT122260-4L BT122260-500ML
Methyl ethyl ketone ≥99.0% ACS
Methyl ethyl ketone

CAS: 78-93-3; EC No: 201-159-0; MDL No: MFCD00011648; RTECS: EL6475000 UN No: UN1193; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8O; MW: 72.11 Melting Point: -87°; Boiling Point: 80°; Flash point: -3°C (27°F) Density (g/mL): 0.805; Refractive Index: 1.379


  • RTECS: EL6475000
  • Refractive index: 1.379
  • Liquid

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: -86
Density: 0.805 g/mL
Flash Pt: –3 °C (27 °F)
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Color (APHA) ≤15
Residue after evaporation ≤25ppm
Distillation range ≤1.5
Water ≤0.2%
Titrable acid ≤0.0005meq/g
Boiling range Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR