2-Chloro-3-fluoropyridine ≥97%
Supplier: BeanTown Chemical
CAS: 17282-04-1; MDL No: MFCD03095302
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C5H3ClFN ; MW: 131.54
Boiling Point: 80°/80 mmHg; Flash point: 63°C (145°F)
Density (g/mL): 1.323; Refractive Index: 1.509
- Refractive index: 1.509
- Liquid
Formula:
C₅H₃ClFN Boiling Pt: 80 °C/80 mmHg Density: 1.323 g/mL Flash Pt: 63 °C (145 °F) |
MDL Number:
MFCD03095302 CAS Number: 17282-04-1 UN: 2810 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...