tert-Butylchlorodimethylsilane ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

122225-100G 122225-25G 122225-5G
BT122225-100GEA 254.6 USD
BT122225-100G BT122225-25G BT122225-5G
tert-Butylchlorodimethylsilane ≥97%
tert-Butylchlorodimethylsilane

CAS: 18162-48-6; EC No: 242-042-4; MDL No: MFCD00000501; RTECS: VV2000000 UN No: UN2925; Haz Class: 4.1 (8); Packing Group: II Crystalline; Linear Formula: (CH3)3CSi(CH3)2Cl; Molecular Formula: C6H15ClSi; MW: 150.73 Melting Point: 86-89°; Boiling Point: 124-126°; Flash point: 22°C (72°F) Density (g/mL): 0.87 Moisture Sensitive


  • RTECS: VV2000000
  • Moisture Sensitive
  • Crystalline

Formula: C₆H₁₅ClSi
MW: 150.72 g/mol
Boiling Pt: 124…126 °C
Melting Pt: 86…89 °C
Density: 0.87 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
ADR: 4.1 (8),II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR