Maleic anhydride ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,5-Furanedione
122195-1KG
122195-250G
122195-5KG
BT122195-1KGEA
32.74
USD
BT122195-1KG
BT122195-250G
BT122195-5KG
Maleic anhydride ≥98%
Maleic anhydride
CAS: 108-31-6; EC No: 203-571-6; MDL No: MFCD00005518; RTECS: ON3675000
UN No: UN2215; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C4H2O3; MW: 98.06
Melting Point: 51-56°; Boiling Point: 200°; Flash point: 103°C (217°F)
Density (g/mL): 1.48
Moisture Sensitive
- RTECS: ON3675000
- Moisture Sensitive
- Solid
Formula:
C₄H₂O₃ MW: 98.06 g/mol Boiling Pt: 200 °C Melting Pt: 51…56 °C Density: 1.48 g/mL Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00005518 CAS Number: 108-31-6 EINECS: 203-571-6 UN: 2215 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...