Sodium cyanotrihydroborate ≥95%
Supplier: BeanTown Chemical
122180-100G
122180-25G
122180-5G
BT122180-100GEA
426.96
USD
BT122180-100G
BT122180-25G
BT122180-5G
Sodium cyanotrihydroborate ≥95%
Sodium cyanotrihydroborate
CAS: 25895-60-7; EC No: 247-317-2; MDL No: MFCD00003516
UN No: UN3134; Haz Class: 4.3 (6.1); Packing Group: II
Powder; Linear Formula: NaBH3CN; MW: 62.84
Melting Point:
- Hygroscopic, Moisture Sensitive
- Powder
Formula:
CH₃BNNa Melting Pt: >242 °C (dec.) Density: 1.2 g/mL |
MDL Number:
MFCD00003516 CAS Number: 25895-60-7 EINECS: 247-317-2 UN: 3134 ADR: 4.3,II |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...